missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4'-methoxyacetophenone, 99%

Product Code. 10351094 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351094 50 g Glass bottle 50g
10439420 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351094 Supplier Thermo Scientific Acros Supplier No. 121330500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 552-41-0 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-41-0
MDL Number MFCD00008730
InChI Key UILPJVPSNHJFIK-UHFFFAOYSA-N
Synonym paeonol, 2'-hydroxy-4'-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethanone, peonol, 2-hydroxy-4-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethan-1-one, resacetophenone-4-methyl ether, ethanone, 1-2-hydroxy-4-methoxyphenyl, paeonal, paeonolum
PubChem CID 11092
ChEBI CHEBI:69581
IUPAC Name 1-(2-hydroxy-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 47°C to 50°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H10O3
Quantity 50 g
Beilstein 08, 267
Molecular Weight (g/mol) 166.18
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Hydroxy-4'-methoxyacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Hydroxy-4'-methoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.