missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%

Product Code. 11490597
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490597 100 g 100g
11470597 5 g 5g
11480597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490597 Supplier Thermo Scientific Alfa Aesar Supplier No. B22445.22
  • ONLINE EXCLUSIVE PRICE
    £785.40
    £641.65 / 100g
    Save £143.75
    18% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2035-72-5 | C17H14O4 | 282.295 g/mol

2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2035-72-5
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00080561
InChI Key IMNBHNRXUAJVQE-UHFFFAOYSA-N
Synonym 4-benzoyl-3-hydroxyphenyl methacrylate, 4-methacryloxy-2-hydroxybenzophenone, 2-hydroxy-4-methacryloyloxy benzophenone, methacrylic acid 4-benzoyl-3-hydroxyphenyl ester, 3-hydroxy-4-phenylcarbonyl phenyl 2-methylprop-2-enoate, 2-propenoic acid, 2-methyl-, 4-benzoyl-3-hydroxyphenyl ester, acmc-20amfp, 2-hydroxy-4-methacryloxybenzophenone, 2-hydroxy-4-methacryloyloxybenzophenone
PubChem CID 74856
IUPAC Name (4-benzoyl-3-hydroxyphenyl) 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 77°C to 79°C
Density 1.18
Quantity 100 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 282.3
Percent Purity 99%
Chemical Name or Material 2-Hydroxy-4-(methacryloyloxy)benzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.