missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-(hydroxymethylphosphoryl) Butanoic acid, TRC

Product Code. 30401893
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401893 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401893 Supplier Toronto Research Chemicals Supplier No. TRCH93978025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-4-(hydroxymethylphosphoryl) Butanoic acid
CAS 80902-10-9
Molecular Formula C5 H11 O5 P
Quantity 25 mg
Synonym Glufosinate Impurity 3
InChI Formula InChI=1S/C5H11O5P/c1-11(9,10)3-2-4(6)5(7)8/h4,6H,2-3H2,1H3,(H,7,8)(H,9,10)
SMILES CP(=O)(O)CCC(O)C(=O)O
IUPAC Name 2-hydroxy-4-[hydroxy(methyl)phosphoryl]butanoic acid
Molecular Weight (g/mol) 182.112
Formula Weight 182.034
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.