missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-benzyloxyacetophenone, TRC

Product Code. 30399405
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399405 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399405 Supplier Toronto Research Chemicals Supplier No. TRCH829860500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-4-benzyloxyacetophenone
CAS 29682-12-0
Molecular Formula C15H14O3
Quantity 500 mg
Synonym Ethanone, 1-[2-hydroxy-4-(phenylmethoxy)phenyl]-, Acetophenone, 4'-(benzyloxy)-2'-hydroxy- (6CI,8CI), 1-[2-Hydroxy-4-(phenylmethoxy)phenyl]ethanone, 1-(4-(Benzyloxy)-2-hydroxyphenyl)ethan-1-one, 1-(4-Benzyloxy-2-hydroxyphenyl)ethanone, 1-[4-(Benzyloxy)-2-hydroxyphenyl]ethan-1-one, 2-Hydroxy-4-(benzyloxy)acetophenone, 2-Hydroxy-4-(phenylmethoxy)acetophenone, 2'-Hydroxy-4'-(benzyloxy)acetophenone, 4-Benzyloxy-2-hydroxy-1-acetophenone, 4-O-Benzylresacetophenone, 4'-(Benzyloxy)-2'-hydroxyacetophenone, 4'-(Benzyloxy)resacetophenone, NSC 211460, Resacetophenone 4-benzyl ether
InChI Formula InChI=1S/C15H14O3/c1-11(16)14-8-7-13(9-15(14)17)18-10-12-5-3-2-4-6-12/h2-9,17H,10H2,1H3
SMILES CC(=O)c1ccc(OCc2ccccc2)cc1O
IUPAC Name 1-(2-hydroxy-4-phenylmethoxyphenyl)ethanone
Molecular Weight (g/mol) 242.27
Formula Weight 242.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.