missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-amino Butanoic Acid, TRC

Product Code. 30391869
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391869 Supplier Toronto Research Chemicals Supplier No. TRCH7972935G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-4-amino Butanoic Acid
CAS 13477-53-7
Molecular Formula C4H9NO3
Quantity 5 g
Synonym 4-Amino-2-hydroxybutanoic Acid, (RS)-2-Hydroxy-GABA, (RS)-2-Hydroxy-gamma-aminobutyric Acid, 2-Hydroxy-gamma-aminobutyric Acid, 4-Amino-2-hydroxybutyric Acid, DL-4-Amino-2-hydroxybutanoic Acid, DL-4-Amino-2-hydroxybutyric Acid, HABAgamma-Amino-alpha-hydroxybutyric Acid
InChI Formula InChI=1S/C4H9NO3/c5-2-1-3(6)4(7)8/h3,6H,1-2,5H2,(H,7,8)
SMILES NCCC(O)C(=O)O
IUPAC Name 4-amino-2-hydroxybutanoic acid
Molecular Weight (g/mol) 119.12
Formula Weight 119.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.