missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%

Product Code. 11411998
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11411998

Brand: Thermo Scientific Alfa Aesar A14585.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-24-4 | C10H12O4 | 196.20 g/mol

2'-Hydroxy-4',6'-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-4′,6′-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-24-4
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00017243
InChI Key FBUBVLUPUDBFME-UHFFFAOYSA-N
Synonym xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
PubChem CID 66654
ChEBI CHEBI:10070
IUPAC Name 1-(2-hydroxy-4,6-dimethoxyphenyl)ethanone
SMILES COC1=CC(O)=C(C(C)=O)C(OC)=C1

Specifications

Melting Point 80°C to 83°C
Boiling Point 185°C (20 mmHg)
Quantity 1 g
Beilstein 1285013
Merck Index 14,10067
Solubility Information Soluble in hot methanol.
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 2'-Hydroxy-4',6'-dimethoxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.