missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4',5'-dimethylacetophenone, 98%, ACROS Organics™

Product Code. 10051740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10051740 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10051740 Supplier Thermo Scientific Acros Supplier No. 156310050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 36436-65-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 69°C to 73°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H12O2
Linear Formula HOC6H2(CH3)2COCH3
MDL Number MFCD00002306
Quantity 5 g
Beilstein 08, II, 122
Synonym 2'-hydroxy-4',5'-dimethylacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethanone, 2-hydroxy-4,5-dimethylacetophenone, 4'5'-dimethyl-2'-hydroxyacetophenone, ethanone, 1-2-hydroxy-4,5-dimethylphenyl, 4',5'-dimethyl-2'-hydroxyacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethan-1-one, 1-acetyl-2-hydroxy-4,5-dimethylbenzene, ethanone,1-2-hydroxy-4,5-dimethylphenyl, acmc-1ctvf
InChI Key YXVSURZEXVMUAM-UHFFFAOYSA-N
SMILES CC1=C(C=C(C(=C1)C(=O)C)O)C
IUPAC Name 1-(2-hydroxy-4,5-dimethylphenyl)ethanone
Molecular Weight (g/mol) 164.204
PubChem CID 118976
Formula Weight 164.2
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2';-Hydroxy-4';, 5';-dimethylacetophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.