missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4,4',6'-trimethoxychalcone, TRC

Product Code. 30393878
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393878 500 mg
30398972 250 mg
30395052 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393878 Supplier Toronto Research Chemicals Supplier No. TRCH214235500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3420-72-2
Molecular Formula C18 H18 O5
Molecular Weight (g/mol) 314.33
Synonym Flavokawain A (cis/trans mixture), Flavokavain A (cis/trans mixture)
IUPAC Name 1-(2-hydroxy-4,6-dimethoxy-phenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
SMILES COc1ccc(C=CC(=O)c2c(O)cc(OC)cc2OC)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H18O5/c1-21-13-7-4-12(5-8-13)6-9-15(19)18-16(20)10-14(22-2)11-17(18)23-3/h4-11,20H,1-3H3
Formula Weight 314.1154
Chemical Name or Material 2'-Hydroxy-4,4',6'-trimethoxychalcone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.