missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Hydroxy-3-N,N,N-trimethylamino)propyl-Gamma-cyclodextrin Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | (2-Hydroxy-3-N,N,N-trimethylamino)propyl-gamma-cyclodextrin Chloride |
| Analyte or Component Names | (2-Hydroxy-3-N,N,N-trimethylamino)propyl-gamma-cyclodextrin Chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C48 H80 nO40 . (C6 H15 O N Cl)n |
| Solution Type | Neat |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 1448.8 |
| Quantity | 500 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?