missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methylpentanoic Acid (Mixture of Diastereomers), TRC

Product Code. 30408762
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408762 100 mg -
30395574 50 mg 50mg
30393737 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408762 Supplier Toronto Research Chemicals Supplier No. TRCH948748100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-15-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.15
Synonym 2-Hydroxy-3-methylvaleric Acid, alpha-Hydroxy-beta-methylvaleric Acid
IUPAC Name 2-hydroxy-3-methylpentanoic acid
SMILES CCC(C)C(O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H12O3/c1-3-4(2)5(7)6(8)9/h4-5,7H,3H2,1-2H3,(H,8,9)
Formula Weight 132.0786
Chemical Name or Material 2-Hydroxy-3-methylpentanoic Acid (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.