missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methylbutyric Acid, TRC

Product Code. 30398127
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398127 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398127 Supplier Toronto Research Chemicals Supplier No. TRCH946510500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-3-methylbutyric Acid
CAS 4026-18-0
Molecular Formula C5 H10 O3
Quantity 500 mg
Synonym Butanoic acid, 2-hydroxy-3-methyl-, Butyric acid, 2-hydroxy-3-methyl- (6CI,7CI,8CI), 2-Hydroxy-3-methylbutanoic acid, (±)-2-Hydroxy-3-methylbutyric acid, (±)-2-Hydroxyisopentanoic acid, (±)-2-Hydroxyisovaleric acid, (±)-α-Hydroxyisovaleric acid, 2-Hydroxy-3-methylbutyric acid, 2-Hydroxyisovaleric acid, 2-Oxyisovaleric acid, 3-Methyl-2-hydroxybutyric acid, DL-2-Hydroxy-3-methylbutanoic acid, DL-2-Hydroxyisovaleric acid, DL-α-Hydroxyisovaleric acid, NSC 227884, dl-2-Hydroxyisovaleric acid, α-Hydroxyisovaleric acid, α-Hydroxyl-3-methylbutyric acid
InChI Formula InChI=1S/C5H10O3/c1-3(2)4(6)5(7)8/h3-4,6H,1-2H3,(H,7,8)
SMILES CC(C)C(O)C(=O)O
IUPAC Name 2-hydroxy-3-methylbutanoic acid
Molecular Weight (g/mol) 118.13
Formula Weight 118.063
Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.