missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-3-methylbutyric Acid-d7, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-3-methylbutyric Acid-d7 |
| Molecular Formula | C5 D7 H3 O3 |
| Quantity | 10 mg |
| Synonym | (±)-2-Hydroxy-3-methylbutyric Acid-d7, (±)-2-Hydroxyisopentanoic Acid-d7, (±)-2-Hydroxyisovaleric Acid-d7, (±)-α-Hydroxyisovaleric Acid-d7, 2-Hydroxy-3-methylbutanoic Acid-d7, 2-Hydroxyisovaleric Acid-d7, 2-Oxyisovaleric Acid-d7, 3-Methyl-2-hydroxybutyric Acid-d7, DL-2-Hydroxy-3-methylbutanoic Acid-d7, dl-2-Hydroxyisovaleric Acid-d7, α-Hydroxyisovaleric Acid-d7 |
| InChI Formula | InChI=1S/C5H10O3/c1-3(2)4(6)5(7)8/h3-4,6H,1-2H3,(H,7,8)/i1D3,2D3,3D |
| SMILES | [2H]C([2H])([2H])C([2H])(C(O)C(=O)O)C([2H])([2H])[2H] |
| IUPAC Name | 3,4,4,4-tetradeuterio-2-hydroxy-3-(trideuteriomethyl)butanoic acid |
| Molecular Weight (g/mol) | 125.174 |
| Formula Weight | 125.1069 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?