missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methylbutanoic acid, MedChemExpress

Product Code. 16479326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479326 Supplier Medchem Express Supplier No. HYW008150
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Hydroxy-3-methylbutanoic acid is a close structure analogue of GHB, which is a naturally occurring neurotransmitter and a psychoactive drug.

2-Hydroxy-3-methylbutanoic acid is a close structure analogue of GHB, which is a naturally occurring neurotransmitter and a psychoactive drug.

Specifications

Chemical Name or Material 2-Hydroxy-3-methylbutanoic acid
CAS 4026-18-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C5H10O3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (846.53 mM; Need ultrasonic)
SMILES CC(C)C(O)C(O)=O
Molecular Weight (g/mol) 118.13
Formula Weight 118.13
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.