missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methoxy-5-nitrobenzaldehyde, 98%

Product Code. p-7021850
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416937 1 g 1g
11426937 5 g 5g
11436937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416937 Supplier Thermo Scientific Alfa Aesar Supplier No. A14360.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17028-61-4 | C8H7NO5 | 197.15 g/mol

2-Hydroxy-3-methoxy-5-nitrobenzaldehyde is used for the syntheses of zinc-selective spiropyran-based fluorescent and photoregenerable receptor, (E)-2,4-dihydroxy-N?-(2-hydroxy-3-methoxy-5-nitrobenzylidene)benzohydrazide dihydrate, ethyl 4-(8-methoxy-3?,3?-dimethyl-6-nitrospiro[chromene-2,2?-indoline]-1?-yl)butanoate and 1-(3-carbomethoxypropyl)-3-3-dimethyl-8-methoxy-6-nitrospiro[2H-1]-benzopyran-2,2-indoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde is used for the syntheses of zinc-selective spiropyran-based fluorescent and photoregenerable receptor, (E)-2,4-dihydroxy-N′-(2-hydroxy-3-methoxy-5-nitrobenzylidene)benzohydrazide dihydrate, ethyl 4-(8-methoxy-3′,3′-dimethyl-6-nitrospiro[chromene-2,2′-indoline]-1′-yl)butanoate and 1-(3-carbomethoxypropyl)-3-3-dimethyl-8-methoxy-6-nitrospiro[2H-1]-benzopyran-2,2-indoline.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17028-61-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
MDL Number MFCD00017033
InChI Key HGKHVFKBOHFYSS-UHFFFAOYSA-N
PubChem CID 307886
IUPAC Name 2-hydroxy-3-methoxy-5-nitrobenzaldehyde
SMILES COC1=CC(=CC(C=O)=C1O)[N+]([O-])=O

Specifications

Melting Point 138°C to 142°C
Quantity 1 g
Beilstein 613271
Sensitivity Air Sensitive
Synonym 3-methoxy-5-nitrosalicylaldehyde, 2-hydroxy-5-nitro-m-anisaldehyde, benzaldehyde, 2-hydroxy-3-methoxy-5-nitro, 5-nitro-o-vanillin, acmc-1bt2m, 3-methoxy-5-nitrosalicyladehyde, 2-hydroxy-5-nitro-3-methoxybenzaldehyde, benzaldehyde,2-hydroxy-3-methoxy-5-nitro
Solubility Information Slightly soluble in water.
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-3-methoxy-5-nitrobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.