missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methoxy-3,3-diphenylpropanoic Acid-d3 Methyl Ester, TRC

Product Code. 30394211
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394211 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394211 Supplier Toronto Research Chemicals Supplier No. TRCH946292100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-3-methoxy-3,3-diphenylpropanoic Acid-d3 Methyl Ester
CAS 1046116-52-2
Molecular Formula C17H15D3O4
Quantity 100 mg
Synonym alpha-Hydroxy-beta-methoxy-beta-phenyl-benzenepropanoic Acid-d3 Methyl Ester
InChI Formula InChI=1S/C17H18O4/c1-20-16(19)15(18)17(21-2,13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12,15,18H,1-2H3/i2D3
SMILES [2H]C([2H])([2H])OC(C(O)C(=O)OC)(c1ccccc1)c2ccccc2
IUPAC Name methyl 2-hydroxy-3,3-diphenyl-3-(trideuteriomethoxy)propanoate
Molecular Weight (g/mol) 289.34
Formula Weight 289.1393
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.