missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-methylpropanamide, TRC

Product Code. 30393730
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393730 500 mg -
30397679 1 g -
30394755 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393730 Supplier Toronto Research Chemicals Supplier No. TRCH513440500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13027-88-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
Synonym 2-Hydroxy-2-methylpropanamide, 2-Hydroxy-2-methylpropionamide, 2-Hydroxyisobutyramide, 2-Methyllactamide, alpha-Hydroxyisobutyramide, alpha-Hydroxyisobutyric Acid Amide
IUPAC Name 2-hydroxy-2-methylpropanamide
SMILES CC(C)(O)C(=O)N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H9NO2/c1-4(2,7)3(5)6/h7H,1-2H3,(H2,5,6)
Formula Weight 103.0633
Chemical Name or Material 2-Hydroxy-2-methylpropanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.