missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-methyl-d3-propanoic-3,3,3-d3 Acid, TRC

Product Code. 30392508
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392508 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392508 Supplier Toronto Research Chemicals Supplier No. TRCH94829210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 40662-45-1
Molecular Formula C4 2H6 H2 O3
Quantity 10 mg
Synonym Propanoic-3,3,3-d3 acid, 2-hydroxy-2-(methyl-d3)- (9CI), 3,3,3-Trideuterio-2-trideuteriomethyl-2-hydroxypropanoic acid, 2-Hydroxy-2-methyl-d3-propanoic-3,3,3-d3 acid, 2-Hydroxy-2-methylpropanoic acid-D6 (dimethyl-D6)
InChI Formula InChI=1S/C4H8O3/c1-4(2,7)3(5)6/h7H,1-2H3,(H,5,6)/i1D3,2D3
SMILES [2H]C([2H])([2H])C(O)(C(=O)O)C([2H])([2H])[2H]
IUPAC Name 3,3,3-trideuterio-2-hydroxy-2-(trideuteriomethyl)propanoic acid
Molecular Weight (g/mol) 110.14
Formula Weight 110.085
Chemical Name or Material 2-Hydroxy-2-methyl-d3-propanoic-3,3,3-d3 Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.