missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-2-benzoylpropane, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-2-methylpropiophenone |
| CAS | 7473-98-5 |
| Molecular Formula | C10 H12 O2 |
| Quantity | 5 g |
| Synonym | 1-Propanone, 2-hydroxy-2-methyl-1-phenyl-, Propiophenone, 2-hydroxy-2-methyl- (6CI,7CI,8CI), 2-Hydroxy-2-methyl-1-phenyl-1-propanone, 1-Hydroxy-1-methylethyl phenyl ketone, 1-Phenyl-2-hydroxy-2-methyl-1-propanone, 1-Phenyl-2-methyl-2-hydroxypropanone, 1173HMPE, 2,2-Dimethyl-2-hydroxyacetophenone, 2-Benzoyl-2-hydroxypropane, 2-Benzoyl-2-propanol, 2-Hydroxy-2,2-dimethylacetophenone, 2-Hydroxy-2-benzoylpropane, 2-Hydroxy-2-methyl-1-phenylpropan-1-one, 2-Hydroxy-2-methyl-1-phenylpropane-1-one, 2-Hydroxy-2-methyl-1-phenylpropanone, 2-Hydroxy-2-methyl-1-propiophenone, 2-Hydroxy-2-methyl-propylphenone, 2-Hydroxy-2-methylphenylpropanone, 2-Hydroxy-2-methylpropiophenone, 2-Hydroxy-methylphenylpropan-1-one, 2-Methyl-2-hydroxy-1-phenyl-1-propanone, 2-Methyl-3-phenyl-3-oxopropan-2-ol, API 1173, Additol HDMAP, AgiSyn 1810, Benacure 1173, Boxing 1173, Chemcure 73, Chivacure 173, Ciba 1173, D 1173, DAR 1173, DC 1173, Darocur 1137, Darocur 1173, Darocur 1173D, Darocur 173, Darocur DR 1173, Doublecure 1173, Doublecure 173, EM 1173, Esacure KL 200, FirstCure HMPP, Gencure 1173, Genocure DHMA, Genocure DMHA, H 0991, HMPP, Hydroxy dimethyl acetophenone, I 1173, IC 1173, IHT-PI 1173, IHT-PI 6022, IHT-PI PBZ, IRG 1173, Irgacure 1173, JRCure 1103, JRcure 1173, MV 5002, Methyl-1-phenylpropan-1-one, Micure HP 8, NSC 401744, Omnirad 1173, Omnirad 73, Photo 1173, Photocure 1173, Photoinitiator 1173, Photoinitiator 1173D, Runtecure 1103, Runtecure 1173, SR 1121, Sarcure 1121, UV 1173, UV 636, α,α-Dimethyl-α-hydroxyacetophenone, α-Hydroxy-α,α-dimethylacetophenone, α-Hydroxy-α-methylpropiophenone, α-Hydroxyisobutyrophenone |
| InChI Formula | InChI=1S/C10H12O2/c1-10(2,12)9(11)8-6-4-3-5-7-8/h3-7,12H,1-2H3 |
| SMILES | CC(C)(O)C(=O)c1ccccc1 |
| IUPAC Name | 2-hydroxy-2-methyl-1-phenylpropan-1-one |
| Molecular Weight (g/mol) | 164.2 |
| Formula Weight | 164.0837 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?