missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-(1-(mercaptomethyl)cyclopropyl)acetic Acid, TRC

Product Code. 30403642
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
10 mg
Unit Size:
0.5mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403642 10 mg 10mg
30399717 2.5 mg 2.5mg
30393245 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403642 Supplier Toronto Research Chemicals Supplier No. TRCH94773010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-2-(1-(mercaptomethyl)cyclopropyl)acetic Acid
Analyte or Component Names 2-Hydroxy-2-(1-(mercaptomethyl)cyclopropyl)acetic Acid
Shipping Condition Room Temperature
Molecular Formula C22 H30 O5
InChI Formula InChI=1S/C22H30O5/c1-6-8-9-12-18(23)13-10-11-16(3)27-22(24)21-17(7-2)14-19(25-4)15-20(21)26-5/h6-7,14-16H,1-2,8-13H2,3-5H3
Solution Type Neat
SMILES COc1cc(OC)c(C(=O)OC(C)CCCC(=O)CCCC=C)c(C=C)c1
IUPAC Name 6-oxoundec-10-en-2-yl 2-ethenyl-4,6-dimethoxybenzoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 374.471
Quantity 10 mg
Formula Weight 374.209
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.