missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-(1-(hydroxymethyl)cyclopropyl)acetonitrile Benzyl Ester, TRC

Product Code. 30398249
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398249 1 g 1g
30405150 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398249 Supplier Toronto Research Chemicals Supplier No. TRCH9465751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-2-(1-(hydroxymethyl)cyclopropyl)acetonitrile Benzyl Ester
Analyte or Component Names 2-Hydroxy-2-(1-(hydroxymethyl)cyclopropyl)acetonitrile Benzyl Ester
Shipping Condition Room Temperature
Molecular Formula C13 H15 N O2
InChI Formula InChI=1S/C13H15NO2/c14-8-12(15)13(6-7-13)10-16-9-11-4-2-1-3-5-11/h1-5,12,15H,6-7,9-10H2
Solution Type Neat
SMILES OC(C#N)C1(COCc2ccccc2)CC1
IUPAC Name 2-hydroxy-2-[1-(phenylmethoxymethyl)cyclopropyl]acetonitrile
Industry Type Pharmaceutical
Molecular Weight (g/mol) 217.264
Quantity 1 g
Formula Weight 217.11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.