missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-2-(1-(hydroxymethyl)cyclopropyl)acetic Acid Benzyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-2-(1-(hydroxymethyl)cyclopropyl)acetic Acid Benzyl Ester |
| Analyte or Component Names | 2-Hydroxy-2-(1-(hydroxymethyl)cyclopropyl)acetic Acid Benzyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H16 O4 |
| InChI Formula | InChI=1S/C13H16O4/c14-11(12(15)16)13(6-7-13)9-17-8-10-4-2-1-3-5-10/h1-5,11,14H,6-9H2,(H,15,16) |
| Solution Type | Neat |
| SMILES | OC(C(=O)O)C1(COCc2ccccc2)CC1 |
| IUPAC Name | 2-hydroxy-2-[1-(phenylmethoxymethyl)cyclopropyl]acetic acid |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 236.264 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?