missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-1-naphthaldehyde, tech.

Product Code. 10421163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10421163 500 g 500g
10347090 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10421163 Supplier Thermo Scientific Acros Supplier No. 121605000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 708-06-5 | C11H8O2 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 708-06-5
MDL Number MFCD00004005
InChI Key NTCCNERMXRIPTR-UHFFFAOYSA-N
Synonym 2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
PubChem CID 12819
IUPAC Name 2-hydroxynaphthalene-1-carbaldehyde
SMILES C1=CC=C2C(=C1)C=CC(=C2C=O)O

Specifications

Melting Point >77.0°C
Color Yellow
Boiling Point 192.0°C (27.0 mmHg)
Infrared Spectrum Authentic
Packaging Plastic bottle
Molecular Formula C11H8O2
Linear Formula HOC10H6CHO
Quantity 500 g
Beilstein 08,143
Solubility Information Solubility in water: 40mg/l
Molecular Weight (g/mol) 172.18
Formula Weight 172.18
Percent Purity ≥93% (on dry substance)
Grade Technical
Physical Form Moist Crystalline Powder
Chemical Name or Material 2-Hydroxy-1-naphthaldehyde, tech.
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-1-naphthaldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.