missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-1-(hydroxymethyl)ethyl 10-undecenoate (>85%), TRC

Product Code. 30392895
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392895 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392895 Supplier Toronto Research Chemicals Supplier No. TRCH578930100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-1-(hydroxymethyl)ethyl 10-undecenoate
CAS 142564-41-8
Molecular Formula C14H26O4
Quantity 100 mg
Synonym 1,3-dihydroxypropan-2-yl undec-10-enoate
InChI Formula InChI=1S/C14H26O4/c1-2-3-4-5-6-7-8-9-10-14(17)18-13(11-15)12-16/h2,13,15-16H,1,3-12H2
SMILES O=C(CCCCCCCCC=C)OC(CO)CO
Molecular Weight (g/mol) 258.36
Formula Weight 258.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.