Learn More
2-Hydroxy-1,4-naphthoquinone, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
2-Hydroxy-1,4-naphthoquinone is used for preparing decorative hair and skin dyes. 2-Hydroxy-1,4-naphthoquinone also demonstrates antimicrobial and antioxidant effects. It also suppress the formation of hydrogen peroxide and superoxide radical anion by aldehyde oxidase-catalyzed reactions.
Solubility
Soluble in water. 2 g/L at 20°C
Notes
Incompatible with strong oxidizing agents. Store at room temperature.
Specifications
Specifications
| Melting Point | ∼193°C (decomposition) |
| Quantity | 250 g |
| Beilstein | 1565260 |
| Merck Index | 14,5393 |
| Solubility Information | Soluble in water. 2g/L at 20°C |
| Formula Weight | 174.16 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 2-Hydroxy-1,4-naphthoquinone |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.