missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinoquinoline, 97%

Product Code. 11410260
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410260 250 mg 250mg
11420260 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410260 Supplier Thermo Scientific Alfa Aesar Supplier No. H50700.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15793-77-8 | C9H9N3 | 159.19 g/mol

2-Hydrazinoquinoline acts as as a derivatization agent for LC-MS-based metabolomic investigation of diabetic ketoacidosis. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydrazinoquinoline acts as as a derivatization agent for LC-MS-based metabolomic investigation of diabetic ketoacidosis. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15793-77-8
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00041860
InChI Key QMVCLSHKMIGEFN-UHFFFAOYSA-N
PubChem CID 85111
IUPAC Name quinolin-2-ylhydrazine
SMILES NNC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 141°C to 143°C
Quantity 250 mg
Synonym 2-hydrazinoquinoline, 2-hydrazinylquinoline, 2-quinolylhydrazine, 2 1h-quinolinone, hydrazone, quinoline, 2-hydrazino, 2-hydrazinequinoline, quinoline, 2-hydrazinyl, quinoline,2-hydrazinyl, .alpha.-quinolylhydrazine, alpha-quinolylhydrazine
Solubility Information Soluble in methanol.
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 2-Hydrazinoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.