Learn More
2-Hydrazinobenzoic acid hydrochloride, 99%, ACROS Organics™
Brand: Acros Organics 411800050
Code : 29
Additional Details : CAS Number : 52356-01-1 Weight : 0.05500kg
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
52356-01-1 | |
188.61 | |
ZGNNOFKURIXXRF-UHFFFAOYSA-N | |
104169 | |
C1=CC=C(C(=C1)C(=O)O)NN.Cl |
C7H8N2O2·HCl | |
MFCD00012931 | |
2-hydrazinobenzoic acid hydrochloride, 2-hydrazinylbenzoic acid hydrochloride, benzoic acid, 2-hydrazino-, monohydrochloride, benzoic acid, 2-hydrazino-, hydrochloride, benzoic acid, 2-hydrazinyl-, hydrochloride 1:1, 2-carboxyphenylhydrazine hydrochloride, 2-hydrazinobenzoicacidhydrochloride, 2-carboxyphenylhydrazinium 1+ chloride, pubchem6411, 2-hydrazinobenzoic acid hydrochloride 1:1 | |
2-hydrazinylbenzoic acid;hydrochloride |
Specifications
2-Hydrazinobenzoic acid hydrochloride | |
Glass bottle | |
175°C | |
99% | |
98.5 | |
98.5% min. (Argentometry) | |
MFCD00012931 | |
ZGNNOFKURIXXRF-UHFFFAOYSA-N | |
2-hydrazinylbenzoic acid;hydrochloride | |
104169 | |
Powder |
Authentic | |
White to Beige | |
5g | |
52356-01-1 | |
100.0 | |
C7H8N2O2·HCl | |
2-hydrazinobenzoic acid hydrochloride, 2-hydrazinylbenzoic acid hydrochloride, benzoic acid, 2-hydrazino-, monohydrochloride, benzoic acid, 2-hydrazino-, hydrochloride, benzoic acid, 2-hydrazinyl-, hydrochloride 1:1, 2-carboxyphenylhydrazine hydrochloride, 2-hydrazinobenzoicacidhydrochloride, 2-carboxyphenylhydrazinium 1+ chloride, pubchem6411, 2-hydrazinobenzoic acid hydrochloride 1:1 | |
C1=CC=C(C(=C1)C(=O)O)NN.Cl | |
188.61 | |
188.61 | |
99% |
Safety and Handling
- 2-Hydrazinobenzoic acid hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.