missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinobenzoic acid hydrochloride, 97%

Product Code. 11313146
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313146 5 g 5g
11323146 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313146 Supplier Thermo Scientific Alfa Aesar Supplier No. L11015.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52356-01-1 | C7H9ClN2O2 | 188.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52356-01-1
Molecular Formula C7H9ClN2O2
Molecular Weight (g/mol) 188.61
MDL Number MFCD00012931
InChI Key ZGNNOFKURIXXRF-UHFFFAOYSA-N
Synonym 2-hydrazinobenzoic acid hydrochloride, 2-hydrazinylbenzoic acid hydrochloride, benzoic acid, 2-hydrazino-, monohydrochloride, benzoic acid, 2-hydrazino-, hydrochloride, benzoic acid, 2-hydrazinyl-, hydrochloride 1:1, 2-carboxyphenylhydrazine hydrochloride, 2-hydrazinobenzoicacidhydrochloride, 2-carboxyphenylhydrazinium 1+ chloride, pubchem6411, 2-hydrazinobenzoic acid hydrochloride 1:1
PubChem CID 104169
IUPAC Name 2-hydrazinylbenzoic acid;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1C(O)=O

Specifications

Melting Point ∽181°C to 183°C (decomposition)
Quantity 5 g
Beilstein 4011728
Formula Weight 188.61
Percent Purity 97%
Chemical Name or Material 2-Hydrazinobenzoic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.