missing translation for 'onlineSavingsMsg'
Learn More

2-Hexynal diethyl acetal, 98%, Thermo Scientific™

Catalog Number 11372366
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
18229-78-2
C10H18O2
170.252
MFCD00041643
VUNHOKCOJYJVBT-UHFFFAOYSA-N
2-hexynal diethyl acetal, 2-hexyne, 1,1-diethoxy, 1,1-diethoxy-2-hexyne, acmc-1c7c9, 2-hexyn-1-al diethyl acetal, 2-hexynyl aldehyde diethyl acetal
87518
1,1-diethoxyhex-2-yne
CCCC#CC(OCC)OCC
This item is not returnable. View return policy
18229-78-2
C10H18O2
170.252
MFCD00041643
VUNHOKCOJYJVBT-UHFFFAOYSA-N
2-hexynal diethyl acetal, 2-hexyne, 1,1-diethoxy, 1,1-diethoxy-2-hexyne, acmc-1c7c9, 2-hexyn-1-al diethyl acetal, 2-hexynyl aldehyde diethyl acetal
87518
1,1-diethoxyhex-2-yne
CCCC#CC(OCC)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.885
Boiling Point 79°C to 80°C (8 mmHg)
Refractive Index 1.431
Quantity 5 g
Beilstein 1703624
Formula Weight 170.25
Percent Purity 98%
Chemical Name or Material 2-Hexynal diethyl acetal
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'