Learn More
2-Hexyn-1-ol, 97%
Description
2-Hexyn-1-ol, is an important organic primary and secondary intermediate. It can be used in Pharmaceuticals, Agrochemicals and Dyestuff industry. It also acts as a chemical solvent in fine chemicals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Hexyn-1-ol, is an important organic primary and secondary intermediate. It can be used in Pharmaceuticals, Agrochemicals and Dyestuff industry. It also acts as a chemical solvent in fine chemicals
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Avoid temperature and direct sunlight.
Specifications
Specifications
| Density | 0.897 |
| Boiling Point | 167°C |
| Flash Point | 68°C (154°F) |
| Linear Formula | CH3(CH2)2C≡CCH2OH |
| Refractive Index | 1.454 |
| Quantity | 5 g |
| Beilstein | 1736274 |
| Solubility Information | It is soluble in water. |
| Formula Weight | 98.15 |
| Percent Purity | 97% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.