missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyl-3-hydroxy-5-[(tetrahydro-2H-pyran-2-yl)oxy]-hexadecanoic Acid, TRC

Product Code. 30393130
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393130 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393130 Supplier Toronto Research Chemicals Supplier No. TRCH2965105MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hexyl-3-hydroxy-5-[(tetrahydro-2H-pyran-2-yl)oxy]-hexadecanoic Acid
CAS 104801-93-6
Molecular Formula C27H52O5
Quantity 5 mg
InChI Formula InChI=1S/C27H52O5/c1-3-5-7-9-10-11-12-13-14-18-23(32-26-20-16-17-21-31-26)22-25(28)24(27(29)30)19-15-8-6-4-2/h23-26,28H,3-22H2,1-2H3,(H,29,30)
SMILES CCCCCCCCCCCC(CC(O)C(CCCCCC)C(=O)O)OC1CCCCO1
IUPAC Name 2-hexyl-3-hydroxy-5-(oxan-2-yloxy)hexadecanoic acid
Molecular Weight (g/mol) 456.7
Formula Weight 456.3815
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.