missing translation for 'onlineSavingsMsg'
Learn More

2-Hexene, cis + trans, tech. 85%

Product Code. 15430328
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430328 25 g 25g
15420328 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430328 Supplier Thermo Scientific Alfa Aesar Supplier No. H53472.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 592-43-8 | C6H12 | 84.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-43-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009473
InChI Key RYPKRALMXUUNKS-HWKANZROSA-N
Synonym cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z
PubChem CID 639661
IUPAC Name (E)-hex-2-ene
SMILES CCCC=CC

Specifications

Density 0.678
Boiling Point 67°C to 69°C
Flash Point −20°C (−4°F)
Linear Formula CH3(CH2)2CH=CHCH3
Refractive Index 1.395
Quantity 25 g
UN Number UN3295
Beilstein 969179
Formula Weight 84.16
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Hexene, Cis + Trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.