missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanoylfuran, 99%

Product Code. 11415996
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415996 25 g 25g
11405996 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415996 Supplier Thermo Scientific Alfa Aesar Supplier No. B22306.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14360-50-0 | C10H14O2 | 166.22 g/mol

2-Hexanoylfuran is used as a food additives

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hexanoylfuran is used as a food additives

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14360-50-0
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00037354
InChI Key YUAYWSBSIJVIBS-UHFFFAOYSA-N
Synonym 2-hexanoylfuran, 2-furyl pentyl ketone, 1-hexanone, 1-2-furanyl, pentyl 2-furyl ketone, 1-2-furyl-1-hexanone, 1-furan-2-yl hexan-1-one, 1-2-furyl hexanone, 2-furyl n-pentyl ketone, 1-2-furanyl-1-hexanone, furan, 2-hexanoyl
PubChem CID 61738
IUPAC Name 1-(furan-2-yl)hexan-1-one
SMILES CCCCCC(=O)C1=CC=CO1

Specifications

Density 0.994
Boiling Point 125°C to 127°C (15 mmHg)
Flash Point 107°C (225°F)
Odor Characteristic
Refractive Index 1.489
Quantity 25 g
Beilstein 2956
Solubility Information Difficult to mix.
Formula Weight 166.22
Percent Purity 99%
Chemical Name or Material 2-Hexanoylfuran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.