missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hexanone, Reagent grade, 98%, Honeywell™
Reagent Grade, 98%
£24.66 - £213.10
Chemical Identifiers
CAS | 591-78-6 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.16 |
MDL Number | MFCD00009482 |
InChI Key | QQZOPKMRPOGIEB-UHFFFAOYSA-N |
Synonym | 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl |
PubChem CID | 11583 |
IUPAC Name | hexan-2-one |
SMILES | CCCCC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15630970
|
Honeywell Research Chemicals
103004-10G |
10 g | Glass Bottle | ||||||
15650970
|
Honeywell Research Chemicals
103004-50G |
50 g | Glass Bottle | ||||||
15640970
|
Honeywell Research Chemicals
103004-250G |
250 g | Glass Bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
591-78-6 | |
100.16 | |
QQZOPKMRPOGIEB-UHFFFAOYSA-N | |
11583 | |
CCCCC(C)=O |
C6H12O | |
MFCD00009482 | |
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl | |
hexan-2-one |
Specifications
591-78-6 | |
Colorless | |
127°C | |
C6H12O | |
n20/D 1.401(lit.) | |
UN1224 | |
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl | |
CCCCC(C)=O | |
100.16 | |
100.16g/mol | |
Reagent | |
2-Hexanone |
-57°C | |
0.812g/mL at 25°C | |
23°C (73.4°F) | |
CH3(CH2)3COCH3 | |
MFCD00009482 | |
1737676 | |
QQZOPKMRPOGIEB-UHFFFAOYSA-N | |
hexan-2-one | |
11583 | |
98% | |
Liquid |
Safety and Handling
P261-P281-P314
H226-H336-H361f-H372
missing translation for 'dotInformation' : Class 3 - PG 3 - Ketones, liquid, n.o.s., HI: all
missing translation for 'einecsNumber' : 209-731-1
missing translation for 'rtecsNumber' : MP1400000