missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, 98%

Product Code. 11442657
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
Unit Size:
100g
10g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442657 500 g 500g
11349609 10 g 10g
11359609 50 g 50g
11432657 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11442657 Supplier Thermo Scientific Alfa Aesar Supplier No. A14867.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-78-6 | C6H12O | 100.16 g/mol

2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (±)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (+/-)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.

Solubility
Miscible with acetone, ethanol and ether. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(C)=O

Specifications

Melting Point -56°C
Density 0.811
Boiling Point 126°C to 127°C
Flash Point 23°C (73°F)
Odor Pungent
Refractive Index 1.401
Quantity 500 g
UN Number UN1224
Beilstein 1737676
Merck Index 14,6033
Solubility Information Miscible with acetone,ethanol and ether. Slightly miscible with water.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material 2-Hexanone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Hexanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Hexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.