missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, 98%

Product Code. 10207593 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10207593 2.5 L Glass bottle 2.5 L
10386830 100 mL Glass bottle 100mL
10021590 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10207593 Supplier Thermo Scientific Acros Supplier No. 146880025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Hexanone, 98%, C6H12O, CAS Number-591-78-6, 2-hexanone, 2-oxohexane, butyl methyl ketone, hexanone, hexanone-2, ketone, butyl methyl, methyl butyl ketone, methyl n-butyl ketone, n-butyl methyl ketone, propylacetone, 500mL, 01, 689, 127 deg.C, 100.0, 97.5, Colorless to Yellow, 0.8100g/mL, 209-731-1 | CAS: 591-78-6 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -57°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 127°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)3COCH3
Refractive Index 1.3995 to 1.4015
Quantity 2.5 L
Beilstein 01, 689
Merck Index 15, 6107
Specific Gravity 0.81
Solubility Information Solubility in water: 20g/l (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Hexanone, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Hexanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H372-Causes damage to organs.
  • H361f-Suspected of damaging fertility.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.