missing translation for 'onlineSavingsMsg'
Learn More

2-(Hexamethyleneimino)ethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11351417
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26487-67-2
Molecular Formula C8H17Cl2N
Molecular Weight (g/mol) 198.131
MDL Number MFCD00012842
InChI Key ZQDSOUPBYJIPNM-UHFFFAOYSA-N
Synonym 1-2-chloroethyl azepane hydrochloride, 2-hexamethyleneimino ethyl chloride hydrochloride, 1h-azepine, hydrochloride, 2-chloroethylhexamethylenemine hydrochloride, 1-2-chloroethyl hexamethyleneimine hydrochloride, 2-chloroethyl hexamethylene imine hydrochloride, 2-chloroethyl azaperhydroepine, chloride, pubchem21389, 1-2-chloroethyl azepane hcl, wln: t7ntj a2g &gh
PubChem CID 24188283
IUPAC Name 1-(2-chloroethyl)azepane;hydrochloride
SMILES C1CCCN(CC1)CCCl.Cl

Specifications

Melting Point 206°C to 212°C
Quantity 100 g
UN Number UN2811
Beilstein 3680388
Sensitivity Hygroscopic
Formula Weight 198.14
Percent Purity 98%
Chemical Name or Material 2-(Hexamethyleneimino)ethyl chloride hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'