missing translation for 'onlineSavingsMsg'
Learn More

2-(Hexamethyleneimino)ethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11341417
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341417 25 g 25g
11331417 5 g 5g
11351417 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11341417 Supplier Thermo Scientific Alfa Aesar Supplier No. L01833.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26487-67-2
Molecular Formula C8H17Cl2N
Molecular Weight (g/mol) 198.131
MDL Number MFCD00012842
InChI Key ZQDSOUPBYJIPNM-UHFFFAOYSA-N
Synonym 1-2-chloroethyl azepane hydrochloride, 2-hexamethyleneimino ethyl chloride hydrochloride, 1h-azepine, hydrochloride, 2-chloroethylhexamethylenemine hydrochloride, 1-2-chloroethyl hexamethyleneimine hydrochloride, 2-chloroethyl hexamethylene imine hydrochloride, 2-chloroethyl azaperhydroepine, chloride, pubchem21389, 1-2-chloroethyl azepane hcl, wln: t7ntj a2g &gh
PubChem CID 24188283
IUPAC Name 1-(2-chloroethyl)azepane;hydrochloride
SMILES C1CCCN(CC1)CCCl.Cl

Specifications

Melting Point 206°C to 212°C
Quantity 25 g
UN Number UN2811
Beilstein 3680388
Sensitivity Hygroscopic
Formula Weight 198.14
Percent Purity 98%
Chemical Name or Material 2-(Hexamethyleneimino)ethyl chloride hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.