missing translation for 'onlineSavingsMsg'
Learn More

2-Hexadecyl-octadecanoic Acid, TRC

Product Code. 30395870
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395870 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395870 Supplier Toronto Research Chemicals Supplier No. TRCH293835250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hexadecyl-octadecanoic Acid
CAS 89547-15-9
Molecular Formula C34 H68 O2
Quantity 250 mg
Synonym 2-Hexadecyloctadecanoic Acid, 2-Palmitylstearic Acid
InChI Formula InChI=1S/C34H68O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33(34(35)36)32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33H,3-32H2,1-2H3,(H,35,36)
SMILES CCCCCCCCCCCCCCCCC(CCCCCCCCCCCCCCCC)C(=O)O
IUPAC Name 2-hexadecyloctadecanoic acid
Molecular Weight (g/mol) 508.9
Formula Weight 508.5219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.