missing translation for 'onlineSavingsMsg'
Learn More

2-Heptylamine, TRC

Product Code. 30137448
Change view
Click to view available options
Quantity:
1 g
50 g
Unit Size:
1g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137448 50 g 50g
17994815 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137448 Supplier Toronto Research Chemicals Supplier No. TRCH28290050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7 H17 N
Molecular Weight (g/mol) 115.22
Synonym 2-Heptanamine, Hexylamine, 1-methyl- (6CI,7CI,8CI), (RS)-2-aminoheptane, (±)-2-Aminoheptane, 1-Methylhexanamine, 1-Methylhexylamine, 2-Aminoheptane, 2-Heptylamine, Armeen L-7, DL-1-Methylhexylamine, Heptamine, Heptamine (sympathomimetic), Heptin, NSC 1091, NSC 27160, Rineptil, Rineptyl, Tuamine, Tuaminoheptan, Tuaminoheptane, dl-2-Aminoheptane
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H17N/c1-3-4-5-6-7(2)8/h7H,3-6,8H2,1-2H3
Formula Weight 115.1361
Chemical Name or Material 2-Heptylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.