missing translation for 'onlineSavingsMsg'
Learn More

2-Heptylamine, 98+%, Thermo Scientific Chemicals

Product Code. 10647953
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7680g/mL
Boiling Point 142.0°C to 144.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(NH2)CH3
Refractive Index 1.4175 to 1.4195
Quantity 500 mL
Beilstein 04,194
Merck Index 15,9982
Specific Gravity 0.768
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol, ether, petroleum ether,, chloroform and benzene
Formula Weight 115.22
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Heptylamine, ≥98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Heptylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'