missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Heptanone, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
100mL
250mL
5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | -35.0°C |
| Color | Colorless |
| Density | 0.8200g/mL |
| Boiling Point | 149.0°C to 150.0°C |
| Flash Point | 39°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Plastic drum |
| Show More |
Safety and Handling
Product Identifier
- 2-Heptanone
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction