missing translation for 'onlineSavingsMsg'
Learn More

2-[[(Guanidino)(imino)methyl]sulfanyl]ethanesulfonic Acid, TRC

Product Code. 30398424
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398424 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398424 Supplier Toronto Research Chemicals Supplier No. TRCG83525050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[((Guanidino)(imino)methyl)sulfanyl]ethanesulfonic Acid
CAS 1391053-66-9
Molecular Formula C4 H10 N4 O3 S2
Quantity 50 mg
Synonym 2-[[[(Aminoiminomethyl)amino]iminomethyl]thio]ethanesulfonic acid, 2-[[(Guanidino)(imino)methyl]sulfanyl]ethanesulfonic Acid, Mesna Imp. B (EP)
InChI Formula InChI=1S/C4H10N4O3S2/c5-3(6)8-4(7)12-1-2-13(9,10)11/h1-2H2,(H,9,10,11)(H5,5,6,7,8)
SMILES NC(=N)NC(=N)SCCS(=O)(=O)O
IUPAC Name 2-(N-carbamimidoylcarbamimidoyl)sulfanylethanesulfonic acid
Molecular Weight (g/mol) 226.28
Formula Weight 226.0194
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.