missing translation for 'onlineSavingsMsg'
Learn More

2-Furoylacetonitrile, 97%

Product Code. 11451838
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451838 25 g 25g
11431838 1 g 1g
11441838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451838 Supplier Thermo Scientific Alfa Aesar Supplier No. A14497.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31909-58-7 | C7H5NO2 | 135.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31909-58-7
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00052210
InChI Key RZNSHBXVTAHWPP-UHFFFAOYSA-N
Synonym 2-furoylacetonitrile, 3-furan-2-yl-3-oxopropanenitrile, 3-2-furyl-3-oxopropanenitrile, 2-cyanoacetylfuran, 2-furanoylacetonitrile, 2-furoyl acetonitrile, pubchem11959, acmc-1cock, 2-2-furanoyl acetonitrile
PubChem CID 141671
IUPAC Name 3-(furan-2-yl)-3-oxopropanenitrile
SMILES O=C(CC#N)C1=CC=CO1

Specifications

Melting Point 77°C to 80°C
Quantity 25 g
Beilstein 111902
Formula Weight 135.12
Percent Purity 97%
Chemical Name or Material 2-Furoylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.