missing translation for 'onlineSavingsMsg'
Learn More

2-Furonitrile, 98+%

Product Code. 11404893
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404893 25 g 25g
11494883 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404893 Supplier Thermo Scientific Alfa Aesar Supplier No. A12214.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-90-3 | C5H3NO | 93.09 g/mol

2-Furonitrile is used as a extractive distillation solvent and as a sweetening agent. It is also used as an intermediate in pharmaceutical and fine chemical synthesis. 2-Furonitrile was employed as substrate to investigate the substrate specificity of nitrilase from Rhodococcus rhodochrous Jl cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Furonitrile is used as a extractive distillation solvent and as a sweetening agent. It is also used as an intermediate in pharmaceutical and fine chemical synthesis. 2-Furonitrile was employed as substrate to investigate the substrate specificity of nitrilase from Rhodococcus rhodochrous Jl cell.

Solubility
Soluble in most common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-90-3
Molecular Formula C5H3NO
Molecular Weight (g/mol) 93.09
MDL Number MFCD00003223
InChI Key YXDXXGXWFJCXEB-UHFFFAOYSA-N
Synonym 2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile
PubChem CID 69245
IUPAC Name furan-2-carbonitrile
SMILES N#CC1=CC=CO1

Specifications

Density 1.062
Boiling Point 146°C to 148°C
Flash Point 35°C (95°F)
Odor Recognizable
Refractive Index 1.479
Quantity 25 g
UN Number UN1992
Beilstein 107033
Solubility Information Soluble in most common organic solvents. Slightly soluble in water.
Formula Weight 93.09
Percent Purity ≥98%
Chemical Name or Material 2-Furonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.