missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic acid hydrazide, 98%

Product Code. 11440204
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440204 50 g 50g
11430204 10 g 10g
11450204 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440204 Supplier Thermo Scientific Alfa Aesar Supplier No. A13630.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3326-71-4 | C5H6N2O2 | 126.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3326-71-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00003235
InChI Key SKTSVWWOAIAIKI-UHFFFAOYSA-N
Synonym 2-furoic acid hydrazide, 2-furoic hydrazide, 2-furohydrazide, furoylhydrazide, 2-furancarbohydrazide, 2-furoylhydrazide, furoic acid, hydrazide, 2-furoic acid, hydrazide, 2-furoylhydrazine, 2-furylcarbonylhydrazide
PubChem CID 18731
IUPAC Name furan-2-carbohydrazide
SMILES C1=COC(=C1)C(=O)NN

Specifications

Melting Point 77°C to 81°C
Quantity 50 g
Beilstein 114435
Formula Weight 126.12
Percent Purity 98%
Chemical Name or Material 2-Furoic acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.