missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic acid, 98%

Product Code. 10309323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10309323 2.5 kg Plastic bottle 2.5kg
10070920 5 g Glass bottle 5g
10497300 100 g Plastic bottle 100g
10754231 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10309323 Supplier Thermo Scientific Acros Supplier No. 119820025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-14-2 | C5H4O3 | 112.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-14-2
MDL Number MFCD00003238
InChI Key SMNDYUVBFMFKNZ-UHFFFAOYSA-N
Synonym 2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
PubChem CID 6919
ChEBI CHEBI:30845
IUPAC Name furan-2-carboxylic acid
SMILES C1=COC(=C1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 129.0°C to 133.0°C
Color White to Beige
Boiling Point 230.0°C to 232.0°C
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C5H4O3
Quantity 2.5 kg
Beilstein 18, 272
Merck Index 15, 4336
Molecular Weight (g/mol) 112.084
Formula Weight 112.08
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Furoic acid, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Furoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.