missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Furfuryl-d5 thiol, CDN
Click to view available options
Quantity:
0.01 g
0.025 g
Unit Size:
0.01g
0.03g
Specifications
Specifications
| Chemical Name or Material | 2-(Mercaptomethyl)furan-D5 |
| Molecular Formula | C5 D5 H O S |
| Quantity | 0.025 g |
| Synonym | 2-Furanmethane-α,α,3,4,5-d5-thiol, α,α,3,4,5-Pentadeutero-2-furfurylthiol, 2-(Mercaptomethyl-d2)furan-3,4,5-d3, 2-(Mercaptomethyl)furan-D5, 2-Furfuryl-d5 thiol |
| InChI Formula | InChI=1 S/C5H6OS/c7-4-5-2-1-3-6-5/h1-3,7 H,4H2/i1D,2 D,3 D,4D2 |
| SMILES | [2 H]c1oc(c([2 H])c1[2 H])C([2 H])([2 H])S |
| IUPAC Name | dideuterio-(3,4,5-trideuteriofuran-2-yl)methanethiol |
| Molecular Weight (g/mol) | 119.1964 |
| Formula Weight | 119.0453 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction