missing translation for 'onlineSavingsMsg'
Learn More

2-Furanone, TRC

Product Code. 30406690
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406690 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406690 Supplier Toronto Research Chemicals Supplier No. TRCF8640505G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 497-23-4
Molecular Formula C4H4O2
Quantity 5 g
Synonym 4-Hydroxycrotonic Acid gamma-Lactone, 2,5-Dihydrofuranone, 2-Buten-4-olide, 2-Oxo-2,5-dihydrofuran, 4-Hydroxy-2-butenoic Acid Lactone, 4-Hydroxy-2-butenoic Acid gamma-Lactone, 5-Oxo-2,5-dihydrofuran-3-yl Ester, 5H-Furan-2-one, Cratone, Isocrotonolactone, NSC 197009, NSC 51296, deltaalpha,beta-Butenolide, alpha,beta-Crotonolactone, gamma-Crotolactone, gamma-Crotonolactone, gamma-Hydroxycrotonic Acid Lactone
InChI Formula InChI=1S/C4H4O2/c5-4-2-1-3-6-4/h1-2H,3H2
SMILES O=C1OCC=C1
IUPAC Name 2H-furan-5-one
Molecular Weight (g/mol) 84.07
Formula Weight 84.0211
Chemical Name or Material 2-Furanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.