missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Furan-2-yl)acetic Acid, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2-(Furan-2-yl)acetic Acid |
| CAS | 2745-26-8 |
| Molecular Formula | C6H6O3 |
| Quantity | 250 mg |
| Synonym | 2-(2-Furyl)acetic acid, 2-Furanylacetic Acid, 2-Furylacetic Acid |
| InChI Formula | InChI=1S/C6H6O3/c7-6(8)4-5-2-1-3-9-5/h1-3H,4H2,(H,7,8) |
| SMILES | OC(=O)Cc1occc1 |
| IUPAC Name | 2-(furan-2-yl)acetic acid |
| Molecular Weight (g/mol) | 126.11 |
| Formula Weight | 126.0317 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction