missing translation for 'onlineSavingsMsg'
Learn More

2-Furaldehyde, 99%

Product Code. 10449500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal drum
Unit Size:
1 L
10 L
2.5 L
250mL
25mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10449500 1 L Glass bottle 1 L
10307940 25 mL Glass bottle 25mL
10686382 250 mL Glass bottle 250mL
10074053 2.5 L Glass bottle 2.5 L
13498418 10 L Metal drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10449500 Supplier Thermo Scientific Acros Supplier No. 181100010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-01-1 | C5H4O2 | 96.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-01-1
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003229
InChI Key HYBBIBNJHNGZAN-UHFFFAOYSA-N
Synonym furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
PubChem CID 7362
ChEBI CHEBI:34768
IUPAC Name furan-2-carbaldehyde
SMILES O=CC1=CC=CO1

Specifications

Acidity 0.02 eq/L max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -37°C
Color Amber to Brown
Density 1.1600g/mL
Boiling Point 159°C to 161°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5250 to 1.527
Quantity 1 L
Residue after Evaporation 0.5% max.
Merck Index 15,4333
Specific Gravity 1.16
Solubility Information Solubility in water: 83g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
Viscosity 1.49 mPa.s (25°C)
Formula Weight 96.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Furaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.